Adenosine free base

Supplier: RPI

Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside

A10050-100.0 A10050-1000.0 A10050-1500.0 A10050-25.0 A10050-5.0 A10050-5000.0 A10050-500.0
76344-486EA 204.4 USD
76344-486 76344-488 76344-490 76344-492 76344-494 76344-496 77510-154
Adenosine free base
Adenosine

Adenosine, Free Base [9-β-D-Ribofuranosyladenine]


Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₀H₁₃N₅O₄
MW: 267.24 g/mol
Melting Pt: 234 …236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005752
CAS Number: 58-61-7
EINECS: 200-389-9
New Product

Order Now

Specification Test Results

Appearance White to off white crystalline powder
Adenine ≤ 0.2 %
Ammonia ≤ 0.0004 %
Chloride (Cl) ≤ 0.007 %
Endotoxin ≤ 100 EU/g

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR