SnapMop™ Flat head Mop Covers, Micronova
Supplier: Micronova
MZSM6AA-9 MZSM6AA-14 MZSM6AA-18 MZSM56-9 MZSM56-9IR MZSM620-9 MZSM620-14 MZSM620-18 MZSM620-14IR MZSM620-18IR MZSM6AW-14 MZSM6AW-18 MZSM11-14NS MZSM11-16IR MZSM6AA-18IR MZSM6AA-9IR MZSM6AW-18IR MZSM6AW-14IR MZSM6AA-14IR
89107-746CS 546.02 USD
89107-746 89107-748 89107-750 89131-856 89131-862 89134-776 89134-778 89134-780 89134-782 89134-784 89134-786 89239-366 52428-588 52428-590 89501-294 10811-248 75845-742 76169-058 76169-060 76342-616 76342-618 76342-620
SnapMop™ Flat head Mop Covers, Micronova
Mops
A range of lightweight, snap-on flat head mop covers with urethane foam interiors, designed for effective cleaning and sanitizing in the cleanroom. A variety of fabric options provide excellent absorbency, effective cleaning, and disinfectant dispersal for specialized cleaning.
  • Easy-to-change covers fit securely over mop frame
  • Urethane foam interiors fully encased in a choice of fabrics
  • NovaPoly: 100% smooth knit polyester provides excellent disinfectant dispersal
  • PolySorb: 100% textured polyester for removing residues and dispersing disinfectant
  • MegaTex:
More Product Information