1 kb Plus DNA Ladder for Safe Stains, New England Biolabs
Supplier: New England Biolabs (NEB)
N0559S
76339-756EA 87 USD
76339-756
1 kb Plus DNA Ladder for Safe Stains, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
The 1 kb Plus DNA ladder for safe stains is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye. It has been optimized for use on GelRed, GelGreen and SYBR Safe precast agarose gels.
  • Optimized for visualization on GelRed, GelGreen and SYBR safe precast agarose gels
  • Convenient, ready-to-load format
  • Size range: 100 bp to 10 kb (100 bp and 1 kb ladder in one!)
  • Easy to identify reference bands
This ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands.