Mesh Basket, Square, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-104A-31
76338-092EA 606.12 USD
76338-092
Mesh Basket, Square, Marlin Steel Wire Products
Baskets Wire Baskets
Wire mesh material handling basket.
  • Electropolished finish
  • With four mesh openings per linear inch
Square design and stainless steel construction are compatible with a variety of equipment and processes. Uses include materials handling, parts degreasing, ultrasonic parts cleaning, and extreme temperature applications.
Order Now
SPECIFICATIONS
Environmentally Preferable