Epredia™ PrintMate™ Slotted Cassettes, Epredia
Supplier: Epredia
B851090BE B851090BL B851090GN B851090GY B851090LA B851090LG B851090LI B851090OR B851090PK B851090TN B851090WH B851090YW B851120BE B851120BL B851120GN B851120GY B851120LA B851120LG B851120LI B851120OR B851120PK B851120TN B851120WH B851120YW
76337-628CS 666.94 USD
76337-628 76337-630 76337-632 76337-634 76337-636 76337-638 76337-640 76337-642 76337-644 76337-646 76337-648 76337-650 76337-700 76337-702 76337-704 76337-706 76337-708 76337-710 76337-712 76337-714 76337-716 76337-718 76337-720 76337-722
Epredia™ PrintMate™ Slotted Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
PrintMate slotted cassettes are designed for ease of use and print clarity.
  • For routine tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Pore style: Slotted
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguardingMore Product Information