Epredia™ PrintMate™ Biopsy Cassettes, Epredia
Supplier: Epredia
B851080BE B851080BL B851080GN B851080GY B851080LA B851080LG B851080LI B851080OR B851080PK B851080TN B851080WH B851080YW B851110BE B851110BL B851110GN B851110GY B851110LA B851110LG B851110LI B851110OR B851110PK B851110TN B851110WH B851110YW
76337-604CS 468.95 USD
76337-604 76337-606 76337-608 76337-610 76337-612 76337-614 76337-616 76337-618 76337-620 76337-622 76337-624 76337-626 76337-676 76337-678 76337-680 76337-682 76337-684 76337-686 76337-688 76337-690 76337-692 76337-694 76337-696 76337-698
Epredia™ PrintMate™ Biopsy Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
PrintMate biopsy cassettes are designed for ease of use and print clarity.
  • For small biopsy tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Pore style: Small squares
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, whileMore Product Information