Epredia™ PrintMate™ Embedding Cassettes in Tubes, Epredia
Supplier: Epredia
B851010BE B851010BL B851010GN B851010GY B851010LA B851010LG B851010LI B851010OR B851010PK B851010TN B851010WH B851010YW B851100BE B851100BL B851100GN B851100GY B851100LA B851100LG B851100LI B851100OR B851100PK B851100TN B851100WH B851100YW
76337-532CS 543.84 USD
76337-532 76337-534 76337-536 76337-538 76337-540 76337-542 76337-544 76337-546 76337-548 76337-550 76337-552 76337-554 76337-652 76337-654 76337-656 76337-658 76337-660 76337-662 76337-664 76337-666 76337-668 76337-670 76337-672 76337-674
Epredia™ PrintMate™ Embedding Cassettes in Tubes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
PrintMate embedding cassettes are designed for ease of use and print clarity.
  • For routine tissue specimens
  • Variety of 12 colors
  • Comes packed in plastic tubes (sleeves) for PrintMate™ Printer compatibility
  • With or without matching lids
  • Pore style: Round holes
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease ofMore Product Information