Epredia™ HistoScreen™ Cassettes, Epredia
Supplier: Epredia
B851000BE B851000BL B851000GN B851000GY B851000LA B851000LG B851000LI B851000OR B851000PK B851000TN B851000WH B851000YW B851001BE B851001BL B851001GN B851001GY B851001LA B851001LG B851001LI B851001OR B851001PK B851001TN B851001WH B851001YW
76337-484CS 1073.49 USD
76337-484 76337-486 76337-488 76337-490 76337-492 76337-494 76337-496 76337-498 76337-500 76337-502 76337-504 76337-506 76337-508 76337-510 76337-512 76337-514 76337-516 76337-518 76337-520 76337-522 76337-524 76337-526 76337-528 76337-530
Epredia™ HistoScreen™ Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
HistoScreen™ cassette mesh screen design accommodates extremely small biopsy specimens.
  • Designed for handling small biopsies and fragile specimens
  • Unique design provides an inner chamber lined with a 'see-through' mesh
  • Eliminates the need for sponges or lens papers
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Variety of 12 colors with matching lids
  • Pore style: Mesh
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
CassetteMore Product Information