ProCulture® Complete Spinner Flask with Angled Sidearms and Magnetically Driven Vortex Impeller, SP Industries
Supplier: SP Industries
BP-3400-104 BP-3400-106 BP-3400-110 BP-3400-112 BP-3400-116
76335-212EA 679.06 USD
76335-212 76335-216 76335-218 76335-220 76335-222 76335-224 76335-226 76335-228
ProCulture® Complete Spinner Flask with Angled Sidearms and Magnetically Driven Vortex Impeller, SP Industries
Flasks Spinner Flasks
Designed to save time and increase cell production, ProCulture® spinner flasks have internal baffles for enhanced agitation and a magnetically driven vortex impeller that provides gentle top-to-bottom stirring.
  • Sidearm openings large enough for 50 ml pipettes
  • Flask base has central bump to prevent material from accumulating at the bottom center
  • Entire assembly made of FDA approved materials
  • Cap made from Polybutylene terephthalate (PBT), red rubber, Ultem®
  • Flask is type 1, class A borosilicate glass construction
  • All components are autoclavable