Water Bath Protective Agent, polyclear MIX 30 PLUS, PolyScience
Supplier: POLYSCIENCE CORP-EQUIP/PARTS
004-300063
76333-932CS 142.5 USD
76333-932
Water Bath Protective Agent, polyclear MIX 30 PLUS, PolyScience
Cleaning Agents Water Bath Protection Reagents
General purpose fluid that prevents algae growth and premature rust formation. Ideal for use in heating baths, circulators, water baths and shaking water baths.
  • For routine applications above 5 °C
  • Easy to use, no dilution required