Empore™ 96-Well Solid Phase Extraction Plates, CDS Analytical
Supplier: CDS ANALYTICAL
6065 70-2007-3988-9 70-2007-3982-2 70-2007-3981-4 70-2007-3983-0
76333-156CS 1005.01 USD
76333-156 76333-158 76333-248 76333-250 76333-252
Empore™ 96-Well Solid Phase Extraction Plates, CDS Analytical
Microplates Solid Phase Extraction Plates (SPE)
Empore SPE 96-well plates improve data and throughput at a low cost.
  • Optimized for smaller sample volumes, which in turn, reduces elution volumes.
  • Eliminate evaporation steps to streamline methods, improve throughput, and reduce labor.
  • Universal compatibility – Standard polypropylene 96-well plate design for automated applications
  • Avoid clogging of frits by fine particulates, which improves LC-MS uptime and column life
  • Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency
The unique matrix of the Empore membrane allows forMore Product Information