Erbium/810 nm Diode LaserShields, NoIR Laser Company
Supplier: NOIR MEDICAL
DRB52 DRB60
76332-552EA 304.37 USD
76332-552 76332-554 76332-556
Erbium/810 nm Diode LaserShields, NoIR Laser Company
Safety Glasses
CE and ANSI certified laser protection eyewear.
  • Lightweight polycarbonate
Available in sporty frame #52, fit-over frame #53, or goggle frame #60.
Certificaciones: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Información para pedidos: Hard-shell case, cleaning cloth, and strap included.