Oakton® pH Buffer Solutions, Cole-Parmer

Supplier: OAKTON INSTRUMENTS MS

WD-00654-00 WD-00654-04 WD-00654-08 00654-07 00654-12 05942-21 00653-07 00653-08 00653-09 00653-10 00653-11 00653-12 00653-13 00654-01 00654-03 05942-22 05942-24 05942-25 05942-41 05942-42 05942-44 05942-45 05942-61 05942-62 05942-64 05942-65 05942-10 05942-11 00653-14
101076-316EA 20.1 USD
101076-316 101076-318 101076-320 76325-290 76325-292 76325-294 76325-296 76325-298 76325-300 76325-302 76325-304 76325-306 76325-308 76325-310 76325-312 76325-314 76325-316 76325-318 76325-320 76325-384 76325-386 76325-388 76325-390 76325-392 76325-394 76325-396 76325-398 76325-400 76325-402
Oakton® pH Buffer Solutions, Cole-Parmer
Buffers pH Calibration Buffers

Ideal for laboratory and field analysis


  • pH buffers are traceable to NIST standard reference materials
  • Buffer solution lets you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
  • pH Buffer Solutions have pH/temperature tables printed on the labels for accurate calibrations (not on 60-mL, but included as insert)
  • Manufactured under ISO 9000 quality guidelines
  • If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days


Envase: Plastic bottles.


Precaución: Store buffers away from direct sunlight.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR