VWR® Disposable Serological Pipettes
Supplier: VWR International
75816-084 75816-086 75816-088 75816-090 75816-092 75816-094 75816-096 75816-098 75816-100 75816-102 75816-104 75816-106 75816-108 75816-110 75816-112 75816-114 75816-116 76184-742 76184-744 76184-746 76184-748 76184-750 76184-752 76184-754 76184-756 76184-872 76201-710 76204-736 76204-738 76320-274 76320-276 76320-278 76320-280 76320-282 76320-284 76320-286 76320-288 76320-290 76501-552 76501-554 76501-556 76501-558 76501-560
75816-084CS 420.95 USD
75816-084 75816-086 75816-088 75816-090 75816-092 75816-094 75816-096 75816-098 75816-100 75816-102 75816-104 75816-106 75816-108 75816-110 75816-112 75816-114 75816-116 76184-742 76184-744 76184-746 76184-748 76184-750 76184-752 76184-754 76184-756 76184-872 76201-710 76204-736 76204-738 76320-274 76320-276 76320-278 76320-280 76320-282 76320-284 76320-286 76320-288 76320-290 76501-552 76501-554 76501-556 76501-558 76501-560
VWR® Disposable Serological Pipettes
Pipettes Serological Pipettes
Polystyrene, transparent and graduated.
  • DNase/RNase-free, non-pyrogenic, non-cytotoxic, non-haemolytic
  • Graduations are calibrated for accurate dispensing to within ±2%
  • Color-coded according to volume
  • Sterile pipettes are sterilized by irradiation
  • Pipettes meet BST/TSE requirements; latex-free
  • Every product is printed with a batch number for traceability
Certificaciones: These disposable pipettes are made of GPPS (general polystyrene), in compliance with USP Class VI. Manufactured in a class 100 000 cleanroom environment. Manufactured under EN ISO 13485More Product Information