PTFE Barrel Pumps, Ultrapure, Burkle
Supplier: BURKLE INC
5606-0600 5606-0601 5606-1000 5606-1001 5606-0400 5606-0401
10217-246EA 1225.18 USD
10217-246 10217-248 10217-250 10217-252 76319-734 76319-736
PTFE Barrel Pumps, Ultrapure, Burkle
Pumps Drum/Transfer Pumps
Due to the high chemical resistance of the medium-contacting parts made of PTFE and FEP, even liquids which attack conventional materials such as PP or PVC can now be safely transferred (except for elemental fluorine) with this barrel pump. The barrel pump is also suitable for transferring ultra-pure liquids. The stopcock prevents hazardous vapors from escaping, and stops dripping after transfer.
  • PTFE, white
  • Excellent chemical resistance
  • Immersion tube Ø32mm (1.26")
  • All valves are made of solid PTFE/PVDF
  • Infinitely variable adjustment of pump immersion
More Product Information