Digital Torque Gauges, SEALS USA INC
Supplier: Nidec Shimpo America
FG-7000T-1 FG-7000T-2 FG-7000T-3
76318-666EA 1762.2 USD
76318-666 76318-668 76318-670
Digital Torque Gauges, SEALS USA INC
Dynamometers
The FG-7000T series digital torque gauges provide testing flexibility with their external torque sensor input. Torque sensors with standard chuck adapter are immediately recognized when connected to the display base. The multiple-language gauge provide menu programming for easy selection and set-up of the instrument to desired requirements.
  • Memory with group categorization to easily separate your tests for review and later download
  • Free software for viewing, graphing and analyzing data on PC
  • Display update rate: 10 times/second
  • Display flashes battery icon
More Product Information