Sapphire Biomolecular Imager, Azure Biosystems
Supplier: Azure Biosystems
IS2011 IS2012
76291-948EA 12745.02 USD
76291-948 76291-950 76291-952 76317-604 76317-606 76317-608 76317-610 76318-248 76318-250 76318-252 76318-254 76318-256
Sapphire Biomolecular Imager, Azure Biosystems
Photodocumentation Systems
The next generation lab imaging platform.
  • With most imaging systems a decision between application flexibility and image quality has to be made
  • Next generation laser scanning system which offers the best of both worlds
  • Unparalleled flexibility with excellent sensitivity and image quality
  • Capture software will deliver high quality quantifiable data
Up to four solid state lasers (488, 520, 658 and 784 nm) offering ultimate excitation sensitivity. Photon multiplier tube (PMT) for fluorescence and phosphor imaging, avalanche photodiodies (APD) for near-More Product Information