HettCube Incubators and Cooled Incubators, Hettich
Supplier: HETTICH INSTRUMENTS
62000-01 62005-01 64000-01 64005-01 66000-01 66005-01
76317-536EA 7996.8 USD
76317-536 76317-538 76317-540 76317-542 76317-544 76317-546
HettCube Incubators and Cooled Incubators, Hettich
Incubators Cooled Incubators
The HettCube range of incubators and cooled incubators provide functionality and customized features.
  • Stainless steel interior
  • Dual control panel and door lock
  • Ø 42 mm bushing on rear panel
  • High-performance insulation
  • Intuitive operation with 4.3" touchscreen
  • Small footprint of only 0.6 m2 for all models
  • USB service interface
  • Easy cleaning
Temperature is primarlily maintained by radiated heat, preventing hotspots forming and cultures drying out. The environmentally friendly water blown foam insulation, together with the gentle air circulationMore Product Information