Sound Recorder/Datalogger, Certified, Sper Scientific
Supplier: Sper Scientific
850018C
76307-710EA 799.43 USD
76307-710
Sound Recorder/Datalogger, Certified, Sper Scientific
Sound Meters
This unique sound meter enables you to playback a live audio recording that is synced with the measured sound intensity graph. It can quickly collect and analyze all standard sound data such as dB levels, LEQ and SPL, but also allows you to pinpoint the cause of various sound levels using the live recording.
  • Accurate within ±1.0 dB this sound meter meets or exceeds all current specs for IEC 61672:2013 class 2 and ANSI S1.4:2014 type 2 meters.
  • Records up to 64000 records complete with date and time stamp that can be stored in the meter for later download
More Product Information