StrepAim® Rapid Dipstick Test, Germaine Laboratories
Supplier: Germaine Laboratories
73025
76304-412KT 92.85 USD
76304-412
StrepAim® Rapid Dipstick Test, Germaine Laboratories
Assays Immunological Assays
StrepAim® is a rapid immunochromatographic assay for the qualitative detection of group A streptococcal antigen directly from throat swabs.