Vesphene® III st Disinfectant Cleaners
Supplier: Steris
1S15CX 1S2008 1S2029
76303-148CS 406.21 USD
76303-148 76303-150 76304-176
Vesphene® III st Disinfectant Cleaners
Cleaning Agents Disinfectants
STERIS' Vesphene® III st Phenolic disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
  • Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy
  • Convenience - Gamma irradiated and double bagged for easy introduction into your aseptic areas
  • Flexibility - Ideal for use in a contamination control rotation
  • Reliability - Complete lot traceability
  • Assurance - Each lot documented by Certificate of Analysis
STERIS' Vesphene® III stMore Product Information