VACUETTE® Blood Collection Needles, Greiner Bio-One
Supplier: Greiner Bio-One
450070 451047 451048 451069 450061 450062
95057-197CS 468.86 USD
95057-197 95057-203 95057-205 95057-353 95057-195 95057-201 95057-207 76302-842 76302-844 76302-848 76359-810 76359-812
VACUETTE® Blood Collection Needles, Greiner Bio-One
Syringe Needles
These needles are ideal for use in routine blood collection procedures.
  • Color-coded for simple identification of needle gauge
  • Designed for single use
  • Single-sample needles for veterinary use only
They are designed exclusively for single use, and feature color-coded protective needle caps to simplify identification of the needle gauge. Pink caps correspond to 18G needles, yellow caps to 20G, green caps to 21G, and black caps to 22G. Available in two lengths, 25.4 or 38.1 mm (1 or 11/2"). Single-sample needles are designed for veterinary use only.
Order Now
SPECIFICATIONS
Environmentally Preferable