Horizon Horizontal Centrifuge, HORIZON 12, Drucker Diagnostics
Supplier: Drucker
00-283-009-000
76300-132EA 2666.53 USD
76300-132
Horizon Horizontal Centrifuge, HORIZON 12, Drucker Diagnostics
Centrifuges Benchtop Centrifuges
The HORIZON 12 routine centrifuge simplifies processing of blood and urine tubes to deliver the best outcome for your patients. Choose from one of three preset cycles, or set and lock to one cycle to prevent accidental changes. The LED lid lighting prevents forgotten tubes and indicates the status of your samples: off when ready, on when running, and flashing when done.
  • Horizontal centrifugation
  • Select from 3 preset cycles
  • LED lid lighting flashes at cycle end
  • Lock to one setting with one button
  • Provides maximum safety and durability
The convectionMore Product Information