Water, deionized

Supplier: HARDY DIAGNOSTICS

Synonyms: Hydrogen oxide

K259 K187
89407-486PK 159.21 USD
89407-486 76299-202
Water, deionized
Water

For the preparation of dilutions and suspensions.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR