Precision™ General Purpose Water Baths with Beads, Thermo Scientific
Supplier: Thermo Fisher Scientific
TSGP28PMO5
76293-204EA 6480 USD
76293-204
Precision™ General Purpose Water Baths with Beads, Thermo Scientific
Baths Bead Baths
Precision general-purpose baths are rugged, high-performance baths ideal for a wide range of lab applications. Capacities range from 2 to 28 l, including shallow and dual-chamber models.
  • Easily operate and monitor bath with icon-based LCD
  • Over-temperature safety circuitry with audible alarm protects work
  • Auto-on and auto-off timers allow you to optimize operation schedules
  • Easily clean the chamber with its seamless stainless steel interior
  • Save valuable bench space with compact baths
  • Protect your work with audible alarms
  • Enhanced hinge design supports open
More Product Information