2’2’-cGAMP ≥98%

Supplier: Cayman Chemical Company

Synonyms: Adenosine-Guanosine 2’,2’-cyclic monophosphate, 2’,2’-Cyclic GMP-AMP, cGAMP(2’-5’), adenylyl-(2’ → 5’)-2’-guanylic acid, cyclic nucleotide

22419-500 22419-1
76293-136EA 506.41 USD
76293-136 76293-138
2’2’-cGAMP ≥98%
2’2’-cGAMP

  • A synthetic dinucleotide


A synthetic dinucleotide containing non-canonical 2'5'-phosphodiester bonds; binds the adapter protein STING (Kd = 287 nM); induces IFN-β production (EC50 = 15.8 nM in L929 cells)

Formula: C₂₀H₂₄N₁₀O₁₃P₂
MW: 674.4 g/mol
Storage Temperature: Freezer
CAS Number: 1465774-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d191fd46fe4139bf9a640b54c871cb343a0ffcc1&vid=d191fd46fe4139bf9a640b54c871cb343a0ffcc1&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d3b6686a3858a63fe7a89c3389292c96983fe1f&vid=0d3b6686a3858a63fe7a89c3389292c96983fe1f&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51b1cef2c88757f722b05106424f863ddbd6ba0c&vid=51b1cef2c88757f722b05106424f863ddbd6ba0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR