Anti-Hapten 4-hydroxy-3-nitrophenyl acetyl Mouse Monoclonal Antibody [clone: B1-8]

Supplier: Enzo Life Sciences
ABS525-0200
76284-880EA 639.83 USD
76284-880
Anti-Hapten 4-hydroxy-3-nitrophenyl acetyl Mouse Monoclonal Antibody [clone: B1-8]
Antibodies
Recombinant monclonal antibody (Mouse IgG3λ).

Recommended Applications: Flow Cytometry, WB

Type: Primary
Antigen: Hapten 4-hydroxy-3-nitrophenyl acetyl
Clonality: Monoclonal
Clone: B1-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3λ
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR