Color Lab Tape
Supplier: GA INTERNATIONAL INC
807656002538 807656002620 807656002651 807656002569 807656002590 807656002682 807656002712 807656002743 807656002774 807656002484 807656002514 807656002606 807656002637 807656002545 807656002576 807656002668 807656002699 807656002729 807656002750 807656002491 807656002521 807656002613 807656002644 807656002552 807656002583 807656002675 807656002705 807656002736 807656002767 807656002507
76275-734RL 24.17 USD
76275-734 76275-736 76275-738 76275-740 76275-742 76275-744 76275-746 76275-748 76275-750 76276-004 76276-006 76276-008 76276-010 76276-012 76276-014 76276-016 76276-018 76276-020 76276-022 76276-024 76276-026 76276-028 76276-030 76276-032 76276-034 76276-036 76276-038 76276-040 76276-042 76276-044
Color Lab Tape
Tapes
Laboratory tape is available in a range of colors, ideal for color-coding. This general use color lab tape can be inscribed for labeling plastic, glass, cardboard, and metal surfaces.
  • Temperature range: −80 to +120 °C (−112 to +248 °F)
  • Available in a wide range of colors and widths, for easy identification and color-coding
  • Ideal for all general use applications in the lab
  • Continuous tape format allows use of as much or little as necessary
  • Resistant to gamma irradiation (tested to up to 50kGy)
Available in an assortment of eye-catching colors andMore Product Information