Smudge and Alcohol-Proof Ribbons
Supplier: GA INTERNATIONAL INC
807656032283 807656032290 807656032306 807656032313 807656999753 807656004792 807656002972 807656004808 807656002989 807656004815 807656002996 807656004884 807656004891 807656003009 807656003016 807656022376 807656003023 807656003030 807656004921 807656004938 807656004945 807656004952 807656004969 807656004914 807656005089 807656005102 807656005119 807656022383 807656005133 807656005140
76210-252RL 71.16 USD
76210-252 76210-254 76210-256 76210-258 76210-456 76274-316 76274-318 76274-322 76274-636 76274-758 76274-760 76274-762 76274-764 76274-766 76274-790 76274-792 76274-794 76274-796 76274-798 76274-802 76274-804 76274-806 76274-808 76274-810 76274-812 76274-814 76274-816 76274-818 76274-866 76274-868 76274-872 76274-876 76274-878 76274-880 76274-882 76274-884 76274-886
Smudge and Alcohol-Proof Ribbons
Printer Ribbons
These resin ribbons for thermal transfer printers will produce a smudge- and alcohol-proof printout when used. Available in a range of sizes, they are compatible with most common thermal transfer printer brands and models.
  • Smudge-proof resin ribbon
  • Standard use
Smudge- and alcohol-proof resin ribbons for thermal transfer printers. Available in either black or white ink formats, the printout obtained when printing with this ribbon will resist spraying and rubbing with alcohols. Range of sizes available, ensuring compatibility with most common barcode printer brands.