Quality Economy Brand Luer Slip Syringes, Air-Tite Products
Supplier: Air-Tite
MSB30 MS1B MSB3 MSB5 MSB10 MSB20
76262-584CS 175.39 USD
76262-584 76262-600 76262-986 76262-988 76262-990 76262-992
Quality Economy Brand Luer Slip Syringes, Air-Tite Products
Syringes General-Purpose Syringes
Quality disposable syringes, made to exacting specifications. Great value at a low cost.
  • Standard three part syringe
  • Latex-free
  • Non-toxic
  • Pyrogen-free
  • Positive plunger stop
Why spend more if you are looking for a basic, functional syringe? The Air-Tite brand syringes are a quality syringe, manufactured under ISO standards. Suitable for laboratory and veterinary use, these basic 3-part syringes offer a lubricated rubber tip plunger for smooth gliding in sterile, individually wrapped packaging.
From 1 to 60 ml in both luer slip and luer lock, theseMore Product Information