Oakton® PCTSTestr™ 5 Waterproof Pocket Tester, Cole-Parmer

Supplier: Antyila Scientific
35634-60
76258-462EA 345.56 USD
76258-462
Oakton® PCTSTestr™ 5 Waterproof Pocket Tester, Cole-Parmer
Electrochemistry Meters pH and Multi-Parameter Meters
Save money by replacing sensor, not tester!

  • Conductivity, pH, salinity, and TDS all in one meter
  • 3-point calibration with automatic recognition of standards
  • Large easy-to-read LCD features two-color backlight
  • Rugged housing is waterproof, dustproof, and floats in water when sensor cap is on
  • Leakproof cap with fill line functions as a sample/calibration and hydrating cup
  • Two years warranty

NIST and USA buffer recognition.

Certificaciones: CE marked, IP 67 rated housing.

Información de suministro: Includes plastic storage case, lanyard, and four AAA batteries.
Order Now

SPECIFICATIONS

Environmentally Preferable
pH Range –2 – 16
pH Resolution 0.01 pH
pH Accuracy ±0.01 pH, ±1 digit
Conductivity Range 0.00 - 20.00 mS/cm
Conductivity Resolution 0.1 µS/cm, 1 µS/cm, 0.01 mS/cm
Conductivity Accuracy ±1% full scale
Salinity Range 0.0 − 10.0 ppt
Salinity Resolution 0.01 ppt
Salinity Accuracy ±1% full scale
TDS Range 0.1 − 10.0 ppt
TDS Resolution 0.1 ppm, 1 ppm, 0.01 ppt
TDS Accuracy ±1% full scale
Temperature Range 0...50 °C (32...122 °F)
Temperature Resolution 0.1 °F/°C
Temperature Accuracy ±0.5 °F/°C
Temperature Compensation Automatic
Display Dual-line LCD
Battery Life 400 hours
Calibration 1 − 3 points

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR