NitriDerm® EP Nitrile Examination Gloves with Extended Cuff, 182 Series, Innovative Healthcare
Supplier: INNOVATIVE HEALTHCARE CORP. TE
182100 182200 182300 182400
76237-901BX 29.76 USD
76237-901 76237-903 76237-905 76239-945
NitriDerm® EP Nitrile Examination Gloves with Extended Cuff, 182 Series, Innovative Healthcare
Gloves
Powder-free nitrile synthetic glove has an extended cuff length to provide extra protection in high risk situations.
  • Excellent donning properties in a soft, low modulus formula
  • Textured, slightly tacky surface for a better grip
  • Thicker and longer than regular exam gloves
  • Non-latex
  • Textured finish
  • No risk of latex allergens
  • Improved wet/dry grip
Certificaciones: Complies with ASTM D6319, ASTM D5151, ASTM F1671.