Mouse EGF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
KOA0157
76237-126EA 760.6 USD
76237-126
Mouse EGF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse EGF AccuSignal ELISA Kit
  • Natural and recombinant mouse EGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: N977-R1029
Epidermal growth factor or EGF is a growth factor that plays an important role in the regulation of cell growth, proliferation, and differentiation by binding to its receptor EGFR. EGF acts by binding with high affinity to epidermal growth factor receptor(EGFR) on the cell surface and stimulating the intrinsic protein-tyrosine kinase activity of the receptor. EGF results in cellular proliferation, differentiation, and survival. It also has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. EGF has strong expression in kidney, salivary gland, cerebrum, and prostate, moderate expression in trachea and thyroid, and low expression in bone marrow, heart, spleen, thymus, uterus, and colon. No expression was detected in adrenal gland, liver, lung, cerebellum, placenta, and small intestine.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml mouse EGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin, EDTA), tissue lysates or urine to each empty well. It is recommended that each mouse EGF standard solution and each sample be measured in duplicate.