Brady® Workstation Scan and Print Basic Kit with BBP12 Printer, Brady Worldwide
Supplier: Brady Worldwide
BWS-SPS-BBP12
76226-002KT 4592.65 USD
76226-002
Brady® Workstation Scan and Print Basic Kit with BBP12 Printer, Brady Worldwide
Printers Label Printers
Create production label workflows using a Brady BBP12 printer and CR1400 barcode scanning device.
  • Scan Mode - Scan existing barcodes to capture data and transfer onto a new label template
  • CSV Mode - Scan the name of a CSV file to print a specified batch of labels
  • Custom Script Mode - Perform advanced label operations such as parse scanned data or serialize labels
  • Scalable Workflows - Expand workflows to additional workstations on large projects using the Export and Import feature
  • Setup as many workflows as needed
Save time and money by maximizingMore Product Information