1-Hydroxycyclohexyl phenyl ketone ≥98.0%

Supplier: TCI America

Synonyms: (1-Hydroxycyclohexyl)(phenyl)methanone

H0617-100G
76220-112EA 86 USD
76220-112
1-Hydroxycyclohexyl phenyl ketone ≥98.0%
1-Hydroxycyclohexyl phenyl ketone

CAS Number: 947-19-3
MDL Number: MFCD00059561
Molecular Formula: C13H16O2
Molecular Weight: 204.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 49
Flash Point (°C): 150


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 1-Benzoylcyclohexanol

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Melting Pt: 49 °C
Flash Pt: 150 °C
MDL Number: MFCD00059561
CAS Number: 947-19-3
EINECS: 213-426-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0d9838914d5968ca030d3cef640c6735be798e7&vid=a0d9838914d5968ca030d3cef640c6735be798e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR