Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Supplier: Simport Scientific
M505-12T M505-8T M505-9T M505-10T M505-11T M505-2T M505-3T M505-4T M505-5T M505-7T
95045-518CS 403.96 USD
95045-518 76214-100 76214-102 76214-104 76214-106 76214-192 76214-194 76214-196 76214-198 76214-202
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Embedding Cassettes Tissue Processing Cassettes
Secure specimens during processing with the Unisette™ Tissue Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevents early separation of base and lid
  • Snap latch and hinge-lock design; cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger ofMore Product Information