Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Supplier: Simport Scientific
M492-2T M492-3T M492-4T M492-5T M492-10T M492-11T M492-12T M492-6T M492-7T M492-8T M492-9T
76214-068CS 895.92 USD
76214-068 76214-070 76214-072 76214-074 76214-076 76214-078 76214-080 76214-180 76214-182 76214-184 76214-186
Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Embedding Cassettes Tissue Processing Cassettes
Secure specimens during processing with the Histosette® II Tissue Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Open and close as often as necessary; cassettes will always relock securely
  • Back-mounted tab allowing an unobstructed anterior marking area
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
More Product Information