Programmable Refrigerant Incubators, Yamato
Supplier: Yamato Scientific
IN-604115V IN-604220V IN-804115V IN-804220V IN-604W115V IN-604W220V IN-804W115V IN-804W220V IN-604WMK115V IN-604WMK220V
76209-618EA 6285.28 USD
76209-618 76209-620 76209-622 76209-624 76209-626 76209-628 76209-630 76209-632 76209-634 76209-636
Programmable Refrigerant Incubators, Yamato
Incubators Cooled Incubators
Standard low temperature forced convection incubators applicable for low temperature tests and environmental tests.
  • Inner glass door keeps temperature stable during sample observation
  • Designed with a large dual glass door and inner door that forms a triplex glass door for improved heat retention (Models IN604W/804W)
  • Interior light for better sample visibility (Models IN604W/804W)
  • Optional slide shaker table available to put in and take out samples easily (Models IN604W/804W)
  • Digital PID Controller
It has an operating temperature range of –10 to+50 °CMore Product Information