Insect Rearing Chambers, Caron Products
Supplier: Caron Products
7340-25-1 7340-25-2 7340-25-3 7340-33-1 7340-33-2 7340-33-3
76205-450EA 30635.34 USD
76205-450 76205-452 76205-454 76205-456 76205-458 76205-460
Insect Rearing Chambers, Caron Products
Testing Chambers Plant Growth Chambers
Caron's 7340-Series Insect Rearing Chambers provide the results users need, without all the usual hassle.
  • Ideal for insects
  • Clean chamber design
  • Facility friendly
Near-ideal rearing conditions, from chambers purpose-built for a challenging and often messy application. Caron provides maximum corrosion protection, with an all-stainless steel chamber, and a state-of-the-art conformal coated refrigeration coil.
The Clean Chamber design makes all interior components, even optional lights, snap out without tools. This ensures simple and straightforwardMore Product Information