DuPont™ Tychem® 2000 SFR Protective Coveralls
Supplier: DuPont
QS127TGRSM000400 QS127TGRMD000400 QS127TGRLG000400 QS127TGRXL000400 QS127TGR2X000400 QS127TGR3X000400 QS127TGR4X000400 QS127TGR5X000400 QS127TGR6X000400 QS127TGR7X000400
76202-702CS 195.32 USD
76202-702 76202-704 76202-706 76202-708 76202-710 76202-712 76202-714 76202-716 76202-718 76202-720
DuPont™ Tychem® 2000 SFR Protective Coveralls
Disposable Garments
Tychem® 2000 SFR Protective Coveralls provide chemical and secondary flame protection in a lightweight garment.
  • Respirator-fit hood lined with ProShield® 6 SFR fabric
  • Covered, braided elastic at hood
  • Nylon zipper with large metal pull
  • Chin flap with double-sided adhesive tape
  • Storm flap covers zipper, which can be sealed by the wearer with double-sided adhesive strip to prevent intrusion at zipper
  • Covered, braided elastic openings at wrist and ankle for tighter fit
  • Taped seams
Tychem® 2000 SFR garments are intended to be worn over primary flameMore Product Information