VWR® ESD Micro Denier Scrub Top with Knit Cuff
Supplier: VWR International
2039ESW1NVYS 2039ESW1NVYM 2039ESW1NVYL 2039ESW1NVY1XL 2039ESW1NVY2XL 2039ESW1NVY3XL 2039ESW1NVY4XL 2039ESW1NVY5XL 2039ESW1NVYXS 2039ESW1NVY6XL
76202-658CS 2271.46 USD
76202-658 76202-660 76202-662 76202-664 76202-666 76202-668 76202-670 76202-672 76202-674 76202-676 76202-678
VWR® ESD Micro Denier Scrub Top with Knit Cuff
Workwear
This style of scrub top is widely used in pharmaceutical cleanroom operations, and meets/exceeds the recommendations outlined in the IEST-RP-CC-003.4, Garment Considerations for Cleanrooms and Other Controlled Environments.
  • Pullover design with three-snap, Henley-style collar that provides added room for easy on/off
  • Binding on collar helps maintain shape and uniformity
  • Raglan-style sleeves provide increased mobility
  • Sleeves include color-coordinated filament knit cuff for extra comfort
  • 5" side slits at garment bottom for added comfort and long-term
More Product Information