Water Bath and Slide Drying Station, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
HRT-FWS1600
76202-622EA 4614.16 USD
76202-622
Water Bath and Slide Drying Station, Springside Scientific
Slide Warmers/Dryers
Two-in-one slide floatation and drying station.
  • Adjustable LED bowl lighting (super bright, max. intensity)
  • On/off timers for both hot plate and bath
  • Magnetic bowl sensor contains no wires to prevent breakage
  • Large 20-slide dryer with adjustable 0 to 100 °C temperature range
  • Will not overheat when dry
  • Full two-year warranty with free loaner
  • Optional on/off operation via cell phone
  • USB and SIM card ports to charge phone or play music
  • Removable extra-deep glass bowl
Unit also includes internal speakers with USB port and headphone jack to play music.