Trio.Bas™ Gas System with MINI Instrument Kit
Supplier: Hardy Diagnostics
BAS654K BAS655K
76196-438EA 18776.31 USD
76196-438 76196-440
Trio.Bas™ Gas System with MINI Instrument Kit
Samplers Air Samplers
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
  • Aspirating head with easy manipulation bayonet closure
  • Manual or automatic operations
  • Ergonomic and balanced design to facilitate handling with ease
  • The air sampler is easily connected to the output of the gas system
  • Each component of the Trio.Gas™ adapter instrument is autoclavable
  • Calibrated regulator guarantees a 100 liters per minute flow rate
The Trio.Bas™ instruments utilize the impaction method of air sampling. The air, passing through an aspiratingMore Product Information