Anti-KRT8 Mouse Monoclonal Antibody [clone: K8/383]

Supplier: Prosci
33-164
76195-550EA 738.11 USD
76195-550
Anti-KRT8 Mouse Monoclonal Antibody [clone: K8/383]
Antibodies
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). Cytokeratin 8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibody that recognizes only cytokeratin 8 and 18. Cytokeratin 8 exists on several types of normal and neoplastic epithelia, including many ductal and glandular epithelia such as colon, stomach, small intestine,More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol KRT8
Antigen Name keratin 8
Antigen Synonyms KRT8
Antibody Type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone K8/383
Reactivity Zebrafish,Human,Rat
Host Mouse
Gene ID P05787
Isotype IgG1
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Immunogen Recombinant human Cytokeratin 8 protein was used as the immunogen for this antibody.
Format Purified Antibody
Form Liquid
Purification Protein G affinity chromatography
Concentration 0.2 mg/mL
Storage Buffer PBS with 0.1 mg/ml BSA and 0.05% sodium azide
Storage Temperature Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
Shipping Temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR