Anti-MFGE8 Mouse Monoclonal Antibody [clone: MFG-06]
Supplier: Prosci
33-436
76194-150EA 738.11 USD
76194-150
Anti-MFGE8 Mouse Monoclonal Antibody [clone: MFG-06]
Antibodies
Recognizes a protein of 40-45kDa, identified as human milk fat globule membrane protein (HMFG), Lactadherin, or MFG-E8. HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.
Western blot: 1-2 ug/ml Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 0.5-1More Product Information