AriaMx Realtime PCR System, Agilent Technologies
Supplier: Agilent Technologies
401490 401491 401492 401493 401494 5190-7702 5190-7708 5190-9370 600906 600907
76193-374PK 283.65 USD
76193-374 76193-376 76193-584 76193-632 76193-634 76193-638 76193-640 76193-644 76193-692 76193-694 76193-696 76193-698 76193-724 76193-726 76193-728 76193-730 76193-732 76193-704 76193-706
AriaMx Realtime PCR System, Agilent Technologies
Thermal Cyclers
The AriaMx Real-time PCR System is a fully integrated quantitative PCR amplification, detection, and data analysis system.
  • Gene expression analysis
  • New genotyping/HRM capability, mRNA quantification
  • NGS quantification: library preparation, result validation, nucleic acid monitoring, rare allele detection
  • Can use self-Installable optical cartridges
  • Operates at a Cq uniformity standard deviation of <0.20 at fast cycling (5 sec at 95 °C / 10 sec at 60 °C)
  • Data acquisition time of <3 sec/plate
  • Dynamic range of 9 orders of magnitude
  • Operates at a maximum
More Product Information