AriaMx Realtime PCR System, Agilent Technologies

Supplier: Agilent Technologies
401490 401491 401492 401493 401494 5190-7702 5190-7708 5190-9370 600906 600907
76193-374PK 283.65 USD
76193-374 76193-376 76193-584 76193-632 76193-634 76193-638 76193-640 76193-644 76193-692 76193-694 76193-696 76193-698 76193-724 76193-726 76193-728 76193-730 76193-732 76193-704 76193-706
AriaMx Realtime PCR System, Agilent Technologies
Thermal Cyclers
The AriaMx Real-time PCR System is a fully integrated quantitative PCR amplification, detection, and data analysis system.

  • Gene expression analysis
  • New genotyping/HRM capability, mRNA quantification
  • NGS quantification: library preparation, result validation, nucleic acid monitoring, rare allele detection
  • Can use self-Installable optical cartridges
  • Operates at a Cq uniformity standard deviation of <0.20 at fast cycling (5 sec at 95 °C / 10 sec at 60 °C)
  • Data acquisition time of <3 sec/plate
  • Dynamic range of 9 orders of magnitude
  • Operates at a maximum
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Compatible With Optical cartridge types: SYBR/FAM, ROX, HEX, CY5, CY3, ATTO 425
Control Accuracy ±0.2 °C
Cooling Rate Average = 2.5 °C/sec or Median = 3.0 °C/sec
Detector Detection sources are 8 filtered photodiodes
Display Resolution = 1024×768 (1280×1024 recommended) pixels
Touch screen interface
Gradient -
Processing Time 3 sec
Sample capacity 10 - 30 µl
Software ARIA software - standard with instrument
Temperature Range 25.0… 99.9 °C
Temperature Uniformity PCR ±0.4 °C
W×D×H 500×460×420 mm
Weight 23 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR