Reverdin Needle, OR Grade, Sklar®
Supplier: Sklar
26-1891 26-1892 26-1893 26-1894
76193-020EA 220.11 USD
76193-020 76193-022 76193-024 76193-026
Reverdin Needle, OR Grade, Sklar®
Dissection Needles
The Reverdin needle is an instrument that resembles a needle holder and needle combination that acts as a suture passer.
  • Made of stainless steel
  • Industry standard
  • Reusable
  • Latex-free
The eye of the Reverdin needle can be adjusted by a sliding mechanism on the handle, thus making it easier to deliver suture material. This assists the user by shortening the difficulty of suturing as well as the time necessary to complete suturing.
Order Now
SPECIFICATIONS
Environmentally Preferable