TC Crile-Wood Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8006 21-8007 21-8008 21-8009 21-8010 21-8011 21-4914 21-8003
89023-316EA 288.67 USD
89023-316 89023-318 89023-320 89023-322 89023-324 89023-326 76192-544 76192-648
TC Crile-Wood Needle Holder, OR Grade, Sklar®
Forceps
TC Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is more delicate than the Mayo-Hegar but less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids inMore Product Information