Econo™ Nail Nipper, Floor Grade, Sklar
Supplier: Sklar
21-472 21-486 21-493 21-482 21-483 21-484 21-497
76192-486BX 303.51 USD
76192-486 76192-540 76192-546 76192-550 76192-552 76192-554 76192-570
Econo™ Nail Nipper, Floor Grade, Sklar
Cutters
The Econo™ Nail Nipper is used for removing portions of the toe nail or the nail plate.
  • Industry standard
  • Made of stainless steel
The Econo™ Nail Nipper features double spring, partially cross-serrated, plier handles with a locking mechanism that is released upon squeezing. This lock is intended to keep the sharp blades closed when the instrument is not being used. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure.